siRNA Details
Gene Symbol: Hist1h1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002321.1 | - | 1153 | |||
AK135614.1 | - | 887 | ||||
AK155502.1 | - | 1158 | ||||
BC089604.1 | - | 1155 | 6763721 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hist1h1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002321.1 | - | 1153 | |||
AK135614.1 | - | 887 | ||||
AK155502.1 | - | 1158 | ||||
BC089604.1 | - | 1155 | 6763721 |