siRNA Details
Gene Symbol: Hist1h1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015787.4 | NP_056602.1 | 1 | 639 | ||
GenBank mRNA | BC089600.1 | - | 639 | 6745595 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hist1h1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015787.4 | NP_056602.1 | 1 | 639 | ||
GenBank mRNA | BC089600.1 | - | 639 | 6745595 |