siRNA Details
| Gene Symbol: Dkk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015814.2 | NP_056629.1 | 8 | 2304 | ||
| GenBank mRNA | AF177400.1 | - | 8 | 2291 | ||
| AJ243964.1 | - | 8 | 2307 | |||
| AK004853.1 | - | 8 | 2304 | |||
| AK013054.2 | - | 7 | 2245 | |||
| BC005519.1 | - | 7 | 2004 | |||
| BC046304.1 | - | 7 | 2219 | 6493718 | ||
| BC050934.1 | - | 7 | 2201 | 6812621 | ||

Dkk3
Genomic Map