siRNA Details
| Gene Symbol: USP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008563.4 | NP_001008563.2 | 18 | 2093 | ||
| NM_001110303.3 | NP_001103773.2 | 18 | 1995 | |||
| NM_006676.7 | NP_006667.3 | 18 | 1995 | |||
| XM_005251665.3 | XP_005251722.1 | 18 | 1983 | |||
| XM_005251666.3 | XP_005251723.1 | 13 | 1557 | |||
| XM_011518161.2 | XP_011516463.1 | 19 | 2335 | |||
| XM_011518162.2 | XP_011516464.1 | 18 | 2514 | |||
| XM_017014221.1 | XP_016869710.1 | 13 | 1557 | |||
| GenBank mRNA | AB023220.1 | - | 17 | 1913 | ||
| AY074877.1 | - | 18 | 2029 | |||
| BC039593.1 | - | 18 | 1939 | 5588842 | ||

USP20
Genomic Map