siRNA Details
| Gene Symbol: Hs6st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015818.2 | NP_056633.2 | 2 | 1784 | ||
| GenBank mRNA | AB024566.1 | - | 2 | 1774 | ||
| AK144809.1 | - | 3 | 1704 | |||
| AK157556.1 | - | 2 | 1648 | |||
| AK165161.1 | - | 1 | 2824 | |||
| AK165385.1 | - | 2 | 1187 | |||
| AK166962.1 | - | 889 | ||||
| AK167151.1 | - | 2 | 1213 | |||
| BC052316.1 | - | 2 | 1784 | 6391204 | ||

Hs6st1
Genomic Map