siRNA Details
| Gene Symbol: Park2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016694.3 | NP_057903.1 | 6 | 764 | ||
| GenBank mRNA | AB019558.1 | - | 6 | 735 | ||
| AF250293.1 | - | 6 | 764 | |||
| AF250294.1 | - | 6 | 767 | |||
| AF250295.1 | - | 6 | 767 | |||
| BC113204.1 | - | 6 | 747 | 40047891 | ||
| FJ555554.1 | - | 4 | 517 | |||
| FJ555556.1 | - | 5 | 497 | |||

Park2
Genomic Map