siRNA Details
| Gene Symbol: Preb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294302.1 | NP_001281231.1 | 3, 4 | 776 | ||
| NM_016703.3 | NP_057912.2 | 3, 4 | 776 | |||
| GenBank mRNA | AF150808.1 | - | 3, 4 | 679 | ||
| AF193017.1 | - | 3, 4 | 688 | |||
| AK013330.1 | - | 3, 4 | 730 | |||
| AK029838.1 | - | 3, 4 | 711 | |||
| AK151288.1 | - | 3, 4 | 758 | |||
| AK213634.1 | - | 1, 2 | 91 | |||
| BC017527.1 | - | 3, 4 | 679 | 4921119 | ||

Preb
Genomic Map