siRNA Details
| Gene Symbol: Hmgn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016710.2 | NP_057919.2 | 4 | 210 | ||
| GenBank mRNA | AB018374.2 | - | 4 | 165 | ||
| AF213454.1 | - | 4 | 190 | |||
| AK013748.1 | - | 3 | 79 | |||
| AK131996.1 | - | 4 | 180 | |||
| AK146755.1 | - | 4 | 186 | |||
| BC021626.1 | - | 4 | 162 | 4219308 | ||
| BC083087.1 | - | 4 | 172 | 6518798 | ||
| BY279928.1 | - | 4 | 235 | |||

Hmgn5
Genomic Map