siRNA Details
| Gene Symbol: Tmod4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016712.3 | NP_057921.1 | 10 | 1157 | ||
| GenBank mRNA | AF177174.1 | - | 9 | 1076 | ||
| AK144408.1 | - | 10 | 1158 | |||
| AK144535.1 | - | 10 | 1158 | |||
| BC068020.1 | - | 9 | 1093 | 30291625 | ||

Tmod4
Genomic Map