siRNA Details
| Gene Symbol: H2afz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016750.3 | NP_058030.1 | 5 | 737 | ||
| GenBank mRNA | AK028176.1 | - | 5 | 634 | ||
| AK088704.1 | - | 5 | 684 | |||
| AK143572.1 | - | 5 | 636 | |||
| AK146181.1 | - | 5 | 669 | |||
| AK151052.1 | - | 5 | 636 | |||
| AK151462.1 | - | 5 | 565 | |||
| AK151509.1 | - | 5 | 636 | |||
| AK152398.1 | - | 5 | 634 | |||
| AK152848.1 | - | 5 | 635 | |||
| AK168178.1 | - | 5 | 636 | |||
| AK169003.1 | - | 5 | 522 | |||
| AK213534.1 | - | 1 | 27 | |||
| AY074806.1 | - | 5 | 634 | |||
| BC079903.1 | - | 5 | 659 | 30622912 | ||
| BC147478.1 | - | 5 | 574 | 9056200 | ||
| BC158035.1 | - | 5 | 574 | 9007338 | ||
| BC171999.1 | - | 5 | 574 | 9054285 | ||
| U70494.1 | - | 5 | 625 | |||
| Gene Symbol: Gm8203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK210116.1 | - | 1 | 171 | ||

H2afz
Genomic Map