siRNA Details
| Gene Symbol: Ech1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016772.1 | NP_058052.1 | 2 | 184 | ||
| GenBank mRNA | AF030343.1 | - | 2 | 184 | ||
| AK088495.1 | - | 2 | 172 | |||
| BC068112.1 | - | 2 | 184 | 5710971 | ||
| BC087924.1 | - | 2 | 245 | 6706361 | ||

Ech1
Genomic Map