siRNA Details
| Gene Symbol: Nasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016777.3 | NP_058057.3 | 7 | 718 | ||
| GenBank mRNA | AF034610.2 | - | 7 | 708 | ||
| AK132690.1 | - | 7 | 697 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nasp| Gene Symbol: Nasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016777.3 | NP_058057.3 | 7 | 718 | ||
| GenBank mRNA | AF034610.2 | - | 7 | 708 | ||
| AK132690.1 | - | 7 | 697 | |||