siRNA Details
| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016784.3 | NP_058064.2 | 3 | 356 | ||
| GenBank mRNA | AF044334.1 | - | 2 | 248 | ||
| AK075993.1 | - | 3 | 344 | |||
| AK076007.1 | - | 3 | 334 | |||
| AK077197.1 | - | 3 | 357 | |||
| AK080068.1 | - | 3 | 363 | |||
| BC006750.1 | - | 3 | 326 | 3709134 | ||
| BC052895.1 | - | 3 | 314 | |||

Plrg1
Genomic Map