siRNA Details
| Gene Symbol: Nptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016789.3 | NP_058069.1 | 3 | 1010 | ||
| GenBank mRNA | AF049124.1 | - | 3 | 900 | ||
| AK134806.1 | - | 3 | 1011 | |||
| BC026054.1 | - | 3 | 1005 | 4038070 | ||
| DV649643.1 | - | 3 | 449 | |||

Nptx2
Genomic Map