siRNA Details
| Gene Symbol: Txnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016792.4 | NP_058072.2 | 3 | 417 | ||
| GenBank mRNA | AF052660.1 | - | 3 | 246 | ||
| AK029807.1 | - | 3 | 418 | |||
| AK051698.1 | - | 3 | 392 | |||
| AK144211.1 | - | 3 | 374 | |||
| AK150326.1 | - | 3 | 380 | |||
| AK150887.1 | - | 3 | 386 | |||
| AK150983.1 | - | 3 | 386 | |||
| AK152101.1 | - | 3 | 386 | |||
| AK167954.1 | - | 3 | 388 | |||
| AK168712.1 | - | 3 | 381 | |||
| BC061123.1 | - | 3 | 444 | 6596299 | ||

Txnl1
Genomic Map