siRNA Details
| Gene Symbol: Pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016798.3 | NP_058078.1 | 7 | 1294 | ||
| GenBank mRNA | AF002283.1 | - | 7 | 1318 | ||
| AK016447.1 | - | 7 | 1271 | |||
| AK078421.1 | - | 7 | 1294 | |||
| BC070418.1 | - | 7 | 1260 | 5714572 | ||

Pdlim3
Genomic Map