siRNA Details
| Gene Symbol: Pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016798.3 | NP_058078.1 | 7 | 1509 | ||
| GenBank mRNA | AK016447.1 | - | 7 | 1486 | ||
| AK078421.1 | - | 7 | 1509 | |||
| BC070418.1 | - | 7 | 1475 | 5714572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdlim3| Gene Symbol: Pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016798.3 | NP_058078.1 | 7 | 1509 | ||
| GenBank mRNA | AK016447.1 | - | 7 | 1486 | ||
| AK078421.1 | - | 7 | 1509 | |||
| BC070418.1 | - | 7 | 1475 | 5714572 | ||