siRNA Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282072.1 | NP_001269001.1 | 12 | 1896 | ||
| NM_001282076.1 | NP_001269005.1 | 13 | 1850 | |||
| NM_016874.3 | NP_058570.1 | 11 | 1726 | |||
| GenBank mRNA | AF102818.1 | - | 11 | 1586 | ||
| AK082115.1 | - | 11 | 1576 | |||
| AK144333.1 | - | 12 | 1665 | |||
| AK146546.1 | - | 11 | 1603 | |||
| CD353366.1 | - | 5 | 662 | |||
| FJ377318.1 | - | 13 | 1710 | |||
| FJ377319.1 | - | 11 | 1571 | |||

Deaf1
Genomic Map