siRNA Details
| Gene Symbol: Ybx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016875.2 | NP_058571.2 | 5 | 763 | ||
| GenBank mRNA | AF073954.1 | - | 5 | 690 | ||
| AF073955.1 | - | 5 | 557 | |||
| AK131786.1 | - | 5 | 741 | |||
| AK161439.1 | - | 1 | 155 | |||
| BC141112.1 | - | 5 | 866 | 9055724 | ||
| DV661016.1 | - | 4 | 365 | |||

Ybx2
Genomic Map