siRNA Details
| Gene Symbol: Eif3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016876.3 | NP_058572.2 | 5 | 286 | ||
| GenBank mRNA | AF108214.1 | - | 5 | 300 | ||
| AK038249.1 | - | 5 | 296 | |||
| AK168362.1 | - | 5 | 289 | |||
| BC008511.1 | - | 5 | 286 | 3592537 | ||

Eif3g
Genomic Map