siRNA Details
| Gene Symbol: Psmd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164177.1 | NP_001157649.1 | 4 | 457 | ||
| NM_016883.4 | NP_058579.2 | 4 | 457 | |||
| GenBank mRNA | AB022022.1 | - | 4 | 413 | ||
| AK009068.1 | - | 4 | 436 | |||
| AK018233.1 | - | 4 | 435 | |||
| AK136400.1 | - | 4 | 436 | |||
| BC026931.1 | - | 4 | 424 | 3987112 | ||
| BC056196.1 | - | 4 | 442 | 30019985 | ||
| CD564300.1 | - | 4 | 424 | |||

Psmd10
Genomic Map