siRNA Details
| Gene Symbol: Psmd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164177.1 | NP_001157649.1 | 5 | 1091 | ||
| NM_016883.4 | NP_058579.2 | 5 | 1170 | |||
| GenBank mRNA | AK018233.1 | - | 5 | 1147 | ||
| AK136400.1 | - | 5 | 1149 | |||
| BC056196.1 | - | 5 | 1155 | 30019985 | ||

Psmd10
Genomic Map