siRNA Details
| Gene Symbol: Sec61a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016906.4 | NP_058602.1 | 12 | 2343 | ||
| GenBank mRNA | AK007696.1 | - | 1 | 705 | ||
| AK077849.1 | - | 1 | 852 | |||
| AK088473.1 | - | 12 | 2344 | |||
| AK153055.1 | - | 12 | 2333 | |||
| AK159349.1 | - | 12 | 2329 | |||
| AK167484.1 | - | 12 | 2272 | |||
| AK200219.1 | - | 1 | 192 | |||
| BC003707.1 | - | 12 | 2269 | 3592772 | ||
| BC006020.1 | - | 8 | 1928 | |||

Sec61a1
Genomic Map