siRNA Details
| Gene Symbol: Tsnax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016909.2 | NP_058605.1 | 6 | 1332 | ||
| GenBank mRNA | AF187040.1 | - | 6 | 1332 | ||
| AK030407.1 | - | 6 | 1262 | |||
| AK146339.1 | - | 6 | 1265 | |||
| AK159223.1 | - | 6 | 1334 | |||
| AK198675.1 | - | 1 | 37 | |||
| BC004611.1 | - | 6 | 1234 | 3585400 | ||

Tsnax
Genomic Map