siRNA Details
| Gene Symbol: Pkd2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163004.1 | NP_001156476.1 | 3 | 279 | ||
| NM_016927.3 | NP_058623.2 | 3 | 279 | |||
| GenBank mRNA | AF182033.1 | - | 3 | 279 | ||
| AK030120.1 | - | 145 | ||||
| AK133423.1 | - | 3 | 276 | |||
| AK136119.1 | - | 383 | ||||
| AK142697.1 | - | 3 | 268 | |||
| AK163272.1 | - | 373 | ||||
| BC138717.1 | - | 3 | 264 | 8861739 | ||

Pkd2l2
Genomic Map