siRNA Details
| Gene Symbol: DNAJB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006736.5 | NP_006727.2 | 9 | 1464 | ||
| GenBank mRNA | AK289626.1 | - | 9 | 1287 | ||
| BC011609.2 | - | 9 | 1406 | 3619734 | ||
| BX478051.1 | - | 1 | 612 | |||
| S37374.1 | - | 8 | 1252 | |||

DNAJB2
Genomic Map