siRNA Details
| Gene Symbol: Cntn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017383.3 | NP_059079.2 | 6 | 947 | ||
| GenBank mRNA | AB032602.1 | - | 5 | 633 | ||
| AK052972.1 | - | 7 | 840 | |||
| BC076594.1 | - | 6 | 947 | 30649661 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntn6| Gene Symbol: Cntn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017383.3 | NP_059079.2 | 6 | 947 | ||
| GenBank mRNA | AB032602.1 | - | 5 | 633 | ||
| AK052972.1 | - | 7 | 840 | |||
| BC076594.1 | - | 6 | 947 | 30649661 | ||