siRNA Details
| Gene Symbol: Ddx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017397.3 | NP_059093.3 | 11 | 1711 | ||
| GenBank mRNA | AF217000.1 | - | 11 | 1586 | ||
| AF220454.1 | - | 11 | 1714 | |||
| AJ250027.1 | - | 11 | 1586 | |||
| AK154518.1 | - | 11 | 1590 | |||
| BC125471.1 | - | 11 | 1711 | 40129986 | ||
| BC137721.1 | - | 11 | 1711 | 8860740 | ||

Ddx20
Genomic Map