siRNA Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017461.2 | NP_059489.2 | 7, 8 | 704 | ||
| GenBank mRNA | BC140998.1 | - | 7, 8 | 705 | 9055610 | |
| BC145081.1 | - | 7, 8 | 705 | 9053629 | ||
| BX523183.1 | - | 20 | ||||
| M37030.1 | - | 7, 8 | 708 | |||

Sept1
Genomic Map