siRNA Details
| Gene Symbol: Zfp316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017467.3 | NP_059495.3 | 7 | 3377 | ||
| GenBank mRNA | AF031955.1 | - | 1 | 3000 | ||
| AF082568.1 | - | 1 | 2996 | |||
| BC057078.1 | - | 7 | 3294 | 5698016 | ||
| BC060099.1 | - | 5 | 2899 | |||

Zfp316
Genomic Map