siRNA Details
Gene Symbol: Pbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017471.2 | NP_059499.1 | 6 | 534 | ||
GenBank mRNA | AF005204.1 | - | 6 | 538 | ||
AK137501.1 | - | 6 | 561 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017471.2 | NP_059499.1 | 6 | 534 | ||
GenBank mRNA | AF005204.1 | - | 6 | 538 | ||
AK137501.1 | - | 6 | 561 |