siRNA Details
| Gene Symbol: Rp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018739.2 | NP_061209.1 | 4 | 467 | ||
| GenBank mRNA | AK146136.1 | - | 4 | 448 | ||
| AK147912.1 | - | 470 | ||||
| AK168391.1 | - | 4 | 460 | |||
| BC048480.1 | - | 4 | 325 | |||
| BC115549.1 | - | 4 | 427 | 40101700 | ||
| BC115550.1 | - | 4 | 427 | 40101701 | ||
| D78255.1 | - | 4 | 446 | |||

Rp9
Genomic Map