siRNA Details
| Gene Symbol: Golga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018748.3 | NP_061218.2 | 24 | 7347 | ||
| GenBank mRNA | AF051357.2 | - | 24 | 7342 | ||
| AK134783.1 | - | 11 | 1992 | |||
| BC007485.1 | - | 10 | 4096 | |||
| BC053000.1 | - | 11 | 4276 | |||

Golga4
Genomic Map