siRNA Details
| Gene Symbol: Nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018757.1 | NP_061227.1 | 6 | 642 | ||
| GenBank mRNA | AF051942.1 | - | 6 | 642 | ||
| AK004201.1 | - | 6 | 551 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nme6| Gene Symbol: Nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018757.1 | NP_061227.1 | 6 | 642 | ||
| GenBank mRNA | AF051942.1 | - | 6 | 642 | ||
| AK004201.1 | - | 6 | 551 | |||