siRNA Details
| Gene Symbol: Ctnnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018761.3 | NP_061231.3 | 2, 3 | 430 | ||
| GenBank mRNA | AF006071.1 | - | 2, 3 | 417 | ||
| AK034393.1 | - | 2, 3 | 387 | |||
| AK082755.1 | - | 2, 3 | 373 | |||
| BC050070.1 | - | 2, 3 | 407 | 30012833 | ||
| CF896873.1 | - | 2, 3 | 357 | |||
| CJ094037.1 | - | 2, 3 | 431 | |||

Ctnnal1
Genomic Map