siRNA Details
| Gene Symbol: Chst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018763.2 | NP_061233.2 | 2 | 2285 | ||
| GenBank mRNA | AB011451.1 | - | 2 | 1737 | ||
| AB011452.1 | - | 2 | 1737 | |||
| AK131820.1 | - | 1 | 365 | |||
| BC051963.1 | - | 2 | 1672 | 5707543 | ||

Chst2
Genomic Map