siRNA Details
| Gene Symbol: UPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006760.3 | NP_006751.1 | 3, 4 | 395 | ||
| GenBank mRNA | AY455312.1 | - | 3, 4 | 342 | ||
| BC113900.1 | - | 3, 4 | 373 | 40029623 | ||
| Y13645.1 | - | 3, 4 | 380 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UPK2| Gene Symbol: UPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006760.3 | NP_006751.1 | 3, 4 | 395 | ||
| GenBank mRNA | AY455312.1 | - | 3, 4 | 342 | ||
| BC113900.1 | - | 3, 4 | 373 | 40029623 | ||
| Y13645.1 | - | 3, 4 | 380 | |||