siRNA Details
| Gene Symbol: Cd160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163496.1 | NP_001156968.1 | 3 | 502 | ||
| NM_001163497.1 | NP_001156969.1 | 3 | 502 | |||
| NM_018767.3 | NP_061237.3 | 3 | 425 | |||
| GenBank mRNA | AF060982.1 | - | 3 | 396 | ||
| AK042093.1 | - | 3 | 426 | |||
| AK139789.1 | - | 3 | 503 | |||
| AK145332.1 | - | 3 | 483 | |||
| BC021596.1 | - | 3 | 397 | 5098004 | ||
| DV660330.1 | - | 3 | 458 | |||

Cd160
Genomic Map