siRNA Details
| Gene Symbol: Dfna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018769.3 | NP_061239.1 | 10 | 1999 | ||
| GenBank mRNA | AF073309.1 | - | 10 | 2000 | ||
| AK016561.1 | - | 2367 | ||||
| AK171278.1 | - | 10 | 1998 | |||
| BC051962.1 | - | 10 | 2058 | |||
| BC086618.1 | - | 10 | 2058 | |||
| BC132303.1 | - | 9 | 1970 | 40130580 | ||

Dfna5
Genomic Map