siRNA Details
| Gene Symbol: Skap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018773.2 | NP_061243.1 | 11 | 1037 | ||
| GenBank mRNA | AB014485.1 | - | 11 | 954 | ||
| AF051324.1 | - | 11 | 952 | |||
| AK076000.1 | - | 11 | 982 | |||
| AK144289.1 | - | 11 | 974 | |||
| AK204408.1 | - | 3 | 173 | |||
| BC003711.1 | - | 11 | 1037 | 3599914 | ||

Skap2
Genomic Map