siRNA Details
| Gene Symbol: Tbc1d8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018775.4 | NP_061245.3 | 7 | 1400 | ||
| GenBank mRNA | AK138630.1 | - | 7 | 1398 | ||
| AK141520.1 | - | 7 | 1389 | |||
| AK141816.1 | - | 7 | 1354 | |||
| AK162371.1 | - | 1 | 175 | |||
| BC005421.1 | - | 4 | 735 | |||
| BC065081.1 | - | 7 | 1334 | 6832495 | ||
| U88873.1 | - | 3 | 620 | |||

Tbc1d8
Genomic Map