siRNA Details
| Gene Symbol: Tbc1d8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018775.4 | NP_061245.3 | 8 | 1448 | ||
| GenBank mRNA | AK138630.1 | - | 8 | 1446 | ||
| AK141520.1 | - | 8 | 1437 | |||
| AK141816.1 | - | 8 | 1402 | |||
| AK162371.1 | - | 2 | 223 | |||
| BC005421.1 | - | 5 | 783 | |||
| BC065081.1 | - | 8 | 1382 | 6832495 | ||
| U88873.1 | - | 4 | 668 | |||

Tbc1d8
Genomic Map