siRNA Details
| Gene Symbol: Foxo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018789.2 | NP_061259.1 | 3 | 1909 | ||
| GenBank mRNA | AB032770.1 | - | 3 | 1689 | ||
| AF114260.1 | - | 3 | 1909 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxo4| Gene Symbol: Foxo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018789.2 | NP_061259.1 | 3 | 1909 | ||
| GenBank mRNA | AB032770.1 | - | 3 | 1689 | ||
| AF114260.1 | - | 3 | 1909 | |||