siRNA Details
| Gene Symbol: KRT38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006771.3 | NP_006762.3 | 7 | 1832 | ||
| GenBank mRNA | AJ786656.1 | - | 7 | 1360 | ||
| BC131715.1 | - | 7 | 1689 | 40123079 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT38| Gene Symbol: KRT38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006771.3 | NP_006762.3 | 7 | 1832 | ||
| GenBank mRNA | AJ786656.1 | - | 7 | 1360 | ||
| BC131715.1 | - | 7 | 1689 | 40123079 | ||