siRNA Details
Gene Symbol: Plxnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018797.2 | NP_061267.1 | 22 | 3754 | ||
GenBank mRNA | AF190578.1 | - | 22 | 3754 | ||
AK033998.1 | - | 4 | 614 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plxnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018797.2 | NP_061267.1 | 22 | 3754 | ||
GenBank mRNA | AF190578.1 | - | 22 | 3754 | ||
AK033998.1 | - | 4 | 614 |