siRNA Details
| Gene Symbol: Pcnx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018814.3 | NP_061284.2 | 14 | 3643 | ||
| GenBank mRNA | AF096286.1 | - | 8 | 1473 | ||
| AK129221.1 | - | 9 | 1523 | |||
| BC057333.1 | - | 7 | 1386 | |||
| BC066849.1 | - | 9 | 1316 | |||
| BC098370.1 | - | 9 | 1233 | |||

Pcnx
Genomic Map