siRNA Details
| Gene Symbol: Nup210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018815.2 | NP_061285.2 | 40 | 6033 | ||
| GenBank mRNA | AF113751.1 | - | 40 | 5999 | ||
| AK078913.1 | - | 3 | 547 | |||
| AK090317.1 | - | 18 | 2816 | |||
| AK173056.1 | - | 40 | 5987 | |||
| BC052468.1 | - | 40 | 5894 | 6835038 | ||

Nup210
Genomic Map