siRNA Details
| Gene Symbol: Snx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110310.1 | NP_001103780.1 | 2, 3 | 394 | ||
| NM_001110311.1 | NP_001103781.1 | 2, 3 | 394 | |||
| NM_018875.2 | NP_061363.2 | 2, 3 | 394 | |||
| GenBank mRNA | AF062484.1 | - | 2, 3 | 321 | ||
| AK011257.1 | - | 2, 3 | 285 | |||
| AK049561.1 | - | 2, 3 | 287 | |||
| AK131838.1 | - | 2, 3 | 384 | |||
| AK146833.1 | - | 2, 3 | 314 | |||
| AK148235.1 | - | 2, 3 | 347 | |||
| AK168604.1 | - | 2, 3 | 285 | |||
| AK169569.1 | - | 2, 3 | 369 | |||
| AK170403.1 | - | 2, 3 | 285 | |||
| AK170992.1 | - | 2, 3 | 381 | |||
| BC002009.1 | - | 2, 3 | 323 | 3592627 | ||
| BY287766.1 | - | 2, 3 | 394 | |||

Snx12
Genomic Map