siRNA Details
| Gene Symbol: Tnfsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019418.3 | NP_062291.1 | 4 | 1758 | ||
| GenBank mRNA | AB029155.1 | - | 4 | 1769 | ||
| BY554892.1 | - | 1 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfsf14| Gene Symbol: Tnfsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019418.3 | NP_062291.1 | 4 | 1758 | ||
| GenBank mRNA | AB029155.1 | - | 4 | 1769 | ||
| BY554892.1 | - | 1 | 310 | |||