siRNA Details
| Gene Symbol: Gnpnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019425.2 | NP_062298.1 | 6 | 1044 | ||
| GenBank mRNA | AJ001006.1 | - | 6 | 1044 | ||
| AK002466.1 | - | 6 | 1065 | |||
| AK006627.1 | - | 1 | 146 | |||
| AK007722.1 | - | 6 | 1049 | |||
| AK007764.1 | - | 6 | 1072 | |||
| AK018499.1 | - | 6 | 1038 | |||
| AK028693.1 | - | 6 | 1071 | |||
| AK050266.1 | - | 6 | 1043 | |||
| AK167870.1 | - | 6 | 1054 | |||
| AK168074.1 | - | 6 | 1054 | |||
| BC031116.1 | - | 6 | 1034 | 5065641 | ||
| BC092047.1 | - | 6 | 1025 | |||

Gnpnat1
Genomic Map